Assay Details
Target Gene Details
Entrez Gene ID: | 18626 |
Gene Name: | period circadian clock 1 |
Gene Aliases: |
Per, m-rigui, mPer1 |
Location: |
Chr.11:69095140-69109965 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Per1 | NM_011065.4 | NP_035195.2 | ||
| AF022992.1 | AAC53355.1 | |||
| AK081813.1 | BAC38339.1 | |||
| AK154900.1 | BAE32911.1 | |||
| BC091645.1 | AAH91645.1 |
More Information
Set Membership: |
|

Genomic Map