Assay Details
Target Gene Details
Entrez Gene ID: | 195046 |
Gene Name: | NLR family, pyrin domain containing 1A |
Gene Aliases: |
CARD7, DEFCAP, Gm14, Gm15, NAC, Nalp1, Nalp1a, Nlrp1, PP1044 |
Location: |
Chr.11:71091197-71144704 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp1a | NM_001004142.2 | NP_001004142.2 | ||
| AY355339.1 | AAR14736.1 | |||
| DQ117601.1 | AAZ40527.1 | |||
| KC539856.1 | ||||
| KC539857.1 | ||||
| KC539858.1 | ||||
| KC539859.1 | ||||
| KC539860.1 | ||||
| KC539861.1 |

Genomic Map