Assay Details
Target Gene Details
Entrez Gene ID: | 18626 |
Gene Name: | period circadian clock 1 |
Gene Aliases: |
Per, m-rigui, mPer1 |
Location: |
Chr.11:69095140-69109965 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Per1 | NM_001159367.1 | 17 | 2261 | NP_001152839.1 |
| NM_011065.4 | 17 | 2276 | NP_035195.2 | |
| AB002108.1 | 16 | 2090 | BAA22634.1 | |
| AF022992.1 | 17 | 2315 | AAC53355.1 | |
| AK081813.1 | 17 | 2274 | BAC38339.1 | |
| AK148202.1 | 17 | 2268 | BAE28412.1 | |
| AK154900.1 | 17 | 2257 | BAE32911.1 | |
| AK172958.1 | 17 | 2261 | BAD32236.1 | |
| BC039768.1 | 16 | 2161 | AAH39768.1 | |
| BC091645.1 | 17 | 2263 | AAH91645.1 |
More Information
Set Membership: |
|

Genomic Map