Assay Details
Target Gene Details
Entrez Gene ID: | 19183 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, ATPase 3, interacting protein |
Gene Aliases: |
C79099, GT198, HOP2, Tbpip |
Location: |
Chr.11:101092141-101095435 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmc3ip | NM_008949.3 | 5 | 471 | NP_032975.1 |
| AB000121.1 | 5 | 436 | BAA23155.1 | |
| AK133462.1 | 5 | 439 | BAE21670.1 | |
| BC030169.1 | 5 | 448 | AAH30169.1 | |
| FJ937966.1 | 5 | 442 | ||
| FJ937967.1 | 4 | 506 |
More Information
Set Membership: |
|

Genomic Map