Assay Details
Target Gene Details
Entrez Gene ID: | 19184 |
Gene Name: | protease (prosome, macropain) 26S subunit, ATPase 5 |
Gene Aliases: |
mSUG1 |
Location: |
Chr.11:106256185-106263112 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmc5 | NM_008950.1 | 7 | 738 | NP_032976.1 |
| AK010505.1 | 7 | 758 | BAB26990.1 | |
| AK012294.1 | 7 | 771 | ||
| AK135451.1 | 7 | 748 | BAE22539.1 | |
| AK145703.1 | 7 | 742 | BAE26599.1 | |
| BC004052.1 | 3 | 433 | AAH04052.1 | |
| BC030840.1 | 7 | 770 | AAH30840.1 | |
| Z54219.1 | 7 | 738 | CAA90961.1 |
More Information
Set Membership: |
|

Genomic Map