Assay Details
Target Gene Details
Entrez Gene ID: | 66599 |
Gene Name: | RAD52 motif 1 |
Gene Aliases: |
2410008M22Rik, AW212028, Rad52b |
Location: |
Chr.11:101627242-101636091 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdm1 | NM_025654.2 | 4 | 459 | NP_079930.1 |
| AK003135.1 | 4 | 455 | BAB22595.1 | |
| AK010457.1 | 4 | 460 | BAB26954.1 | |
| AK169668.1 | 3 | 1319 | ||
| BC026811.1 | 4 | 439 | AAH26811.1 | |
| BC055013.1 | 4 | 448 | AAH55013.1 |

Genomic Map