Assay Details
Target Gene Details
Entrez Gene ID: | 319454 |
Gene Name: | regulatory associated protein of MTOR, complex 1, opposite strand |
Gene Aliases: |
A430071A18Rik |
Location: |
Chr.11:119820811-119834365 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rptoros | NR_045312.1 | 4 | 398 | |
NR_045313.1 | 3 | 283 | ||
AK040159.1 | 4 | 390 | ||
AK086908.1 | 3 | 284 |
