Assay Details
Target Gene Details
Entrez Gene ID: | 237979 |
Gene Name: | sidekick homolog 2 (chicken) |
Gene Aliases: |
4632412F08Rik, 5330435L01Rik, Sdk-2, mKIAA1514 |
Location: |
Chr.11:113776374-114066181 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdk2 | NM_172800.2 | 27 | 3849 | NP_766388.2 |
| AK129379.1 | 23 | 3230 | BAC98189.1 | |
| AY351699.1 | 27 | 3849 | AAQ57661.1 |

Genomic Map