Assay Details
Target Gene Details
Entrez Gene ID: | 80879 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 3 |
Gene Aliases: |
Mct3, Mct4 |
Location: |
Chr.11:120948484-120959000 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc16a3 | NM_030696.3 | NP_109621.1 | ||
| BC046525.1 | AAH46525.1 |
More Information
Set Membership: |
|

Genomic Map