Assay Details
Target Gene Details
Entrez Gene ID: | 67811 |
Gene Name: | polymerase (DNA-directed), delta interacting protein 2 |
Gene Aliases: |
1300003F06Rik, Pdip38 |
Location: |
Chr.11:78512296-78522736 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Poldip2 | NM_026389.3 | NP_080665.1 | ||
AK003702.1 | ||||
AK165703.1 | ||||
AK178603.1 | ||||
AK183751.1 | ||||
AK192438.1 | ||||
AK201477.1 | ||||
AK206291.1 | ||||
AK218854.1 | ||||
BC006833.1 | AAH06833.1 | |||
BC012879.1 | AAH12879.1 |
