Assay Details
Target Gene Details
Entrez Gene ID: | 237979 |
Gene Name: | sidekick homolog 2 (chicken) |
Gene Aliases: |
4632412F08Rik, 5330435L01Rik, Sdk-2, mKIAA1514 |
Location: |
Chr.11:113776374-114066181 on Build GRCm38 |
Assay Gene Location: | Within Exon 42 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sdk2 | NM_172800.2 | 42 | 5924 | NP_766388.2 |
AK129379.1 | 38 | 5305 | BAC98189.1 | |
AK161326.1 | 10 | 1290 | BAE36324.1 | |
AY351699.1 | 42 | 5924 | AAQ57661.1 |
