Assay Details
Target Gene Details
Entrez Gene ID: | 67465 |
Gene Name: | splicing factor 3a, subunit 1 |
Gene Aliases: |
1200014H24Rik, 5930416L09Rik, AI159724 |
Location: |
Chr.11:4160354-4182541 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sf3a1 | NM_026175.5 | 6 | 972 | NP_080451.4 |
| AK004763.1 | 6 | 955 | ||
| AK028829.1 | 6 | 924 | BAC26142.1 | |
| AK029095.1 | 6 | 942 | BAC26294.1 | |
| AK030223.1 | 6 | 959 | BAC26853.1 | |
| AK134178.1 | 6 | 951 | ||
| AK160061.1 | 6 | 955 | BAE35597.1 | |
| AK215839.1 | 3 | 327 | ||
| BC029753.1 | 6 | 936 | AAH29753.1 |
More Information
Set Membership: |
|

Genomic Map