Assay Details
Target Gene Details
Entrez Gene ID: | 18102 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 1 |
Gene Aliases: |
AL024257, NDPK-A, NM23-M1, NM23A |
Location: |
Chr.11:93958925-93968521 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nme1 | NM_008704.2 | NP_032730.1 | ||
| AF033377.1 | AAB87689.1 | |||
| AK136353.1 | BAE22943.1 | |||
| BC005629.1 | AAH05629.1 | |||
| BY037001.1 | ||||
| M35970.1 | AAA39826.1 | |||
| M65037.1 | AAA63391.1 | |||
| U85511.1 | AAB42080.1 |
More Information
Set Membership: |
|

Genomic Map