Assay Details
Target Gene Details
Entrez Gene ID: | 72129 |
Gene Name: | peroxisomal biogenesis factor 13 |
Gene Aliases: |
2610008O20Rik |
Location: |
Chr.11:23646479-23665935 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pex13 | NM_023651.4 | 1 | 59 | NP_076140.2 |
| AF329877.2 | AAK15313.2 | |||
| AJ304506.1 | CAC20705.1 | |||
| AK011355.1 | BAB27563.1 | |||
| AK029099.1 | BAC26296.1 | |||
| AK029336.1 | BAC26402.1 | |||
| AK031710.1 | 1 | 59 | BAC27526.1 | |
| AK031975.1 | BAC27635.1 | |||
| AK087918.1 | ||||
| AK153438.1 | BAE31995.1 | |||
| AK169148.1 | 1 | 84 | BAE40927.1 | |
| BC023683.1 | 1 | 71 | AAH23683.1 |
Target Gene Details
Entrez Gene ID: | 74467 |
Gene Name: | pseudouridylate synthase 10 |
Gene Aliases: |
2810013G11Rik, 4933435A13Rik, AU014648, C77560, Ccdc139 |
Location: |
Chr.11:23665354-23732876 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pus10 | NM_001033654.2 | NP_001028826.1 | ||
| NM_028956.4 | NP_083232.1 | |||
| AK017063.1 | BAB30576.1 | |||
| AK029866.1 | BAC26650.1 |
More Information
Set Membership: |
|

Genomic Map