Assay Details
Target Gene Details
Entrez Gene ID: | 327958 |
Gene Name: | PITPNM family member 3 |
Gene Aliases: |
A330068P14Rik, AI848332, Ackr6, Gm880 |
Location: |
Chr.11:72047528-72135889 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitpnm3 | NM_001024927.2 | 18 | 2609 | NP_001020098.1 |
| NM_001081641.1 | 17 | 2561 | NP_001075110.1 | |
| AK147444.1 | 18 | 2566 | BAE27916.1 | |
| AK147630.1 | 17 | 2394 | BAE28035.1 | |
| BC112411.1 | 14 | 2523 | ||
| CF531644.1 |
More Information
Set Membership: |
|

Genomic Map