Assay Details
Target Gene Details
Entrez Gene ID: | 78887 |
Gene Name: | Sfi1 homolog, spindle assembly associated (yeast) |
Gene Aliases: |
- |
Location: |
Chr.11:3131850-3203343 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sfi1 | NM_030207.2 | 2 | 164 | NP_084483.2 |
| AK046135.1 | 2 | 161 | ||
| AK046373.1 | 2 | 161 | ||
| AK082866.1 | 2 | 126 | ||
| AK129159.1 | 2 | 131 | BAC97969.1 | |
| AK133558.1 | 2 | 112 | BAE21725.1 | |
| AK170090.1 | 2 | 124 | ||
| BC046305.1 | 2 | 112 | AAH46305.1 | |
| BY303229.1 | 2 | 165 |

Genomic Map