Assay Details
Target Gene Details
Entrez Gene ID: | 18103 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 2 |
Gene Aliases: |
NM23-H2, NM23B, nm23-M2 |
Location: |
Chr.11:93949814-93956256 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nme2 | NM_001077529.2 | NP_001070997.1 | ||
| NM_008705.5 | NP_032731.1 | |||
| AK012447.1 | BAB28246.1 | |||
| AK150419.1 | BAE29543.1 | |||
| AK151262.1 | BAE30250.1 | |||
| AK169120.1 | BAE40900.1 | |||
| AK210541.1 | ||||
| AK216511.1 | ||||
| AV458476.1 | ||||
| BC066995.1 | AAH66995.1 | |||
| BC086892.1 | AAH86892.1 | |||
| BC086893.1 | AAH86893.1 | |||
| BY274368.1 | ||||
| EF437324.1 | ||||
| X68193.1 | CAA48275.1 |
More Information
Set Membership: |
|

Genomic Map