Assay Details
Target Gene Details
Entrez Gene ID: | 78887 |
Gene Name: | Sfi1 homolog, spindle assembly associated (yeast) |
Gene Aliases: |
- |
Location: |
Chr.11:3131850-3203343 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sfi1 | NM_030207.2 | NP_084483.2 | ||
| AK046135.1 | ||||
| AK046373.1 | ||||
| AK129159.1 | BAC97969.1 | |||
| AK133558.1 | BAE21725.1 | |||
| AK170090.1 | ||||
| BC046305.1 | AAH46305.1 |

Genomic Map