Assay Details
Target Gene Details
Entrez Gene ID: | 15567 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter, serotonin), member 4 |
Gene Aliases: |
5-HTT, AI323329, Htt, Sert |
Location: |
Chr.11:76998466-77032345 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a4 | NM_010484.2 | 13 | 1928 | NP_034614.2 |
| AF013604.1 | 12 | 1741 | AAB67172.1 | |
| AK008410.1 | 2 | 129 | ||
| AY765218.1 | 2 | 186 | AAV41217.1 | |
| BC108978.1 | 13 | 1910 | AAI08979.1 | |
| BC108979.1 | 13 | 1910 | AAI08980.1 | |
| X66119.1 | 12 | 1522 |
More Information
Set Membership: |
|

Genomic Map