Assay Details
Target Gene Details
Entrez Gene ID: | 320655 |
Gene Name: | post-GPI attachment to proteins 3 |
Gene Aliases: |
CAB2, D430035D22Rik, Perld1 |
Location: |
Chr.11:98388672-98400539 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pgap3 | NM_001033537.2 | 3 | 412 | NP_001028709.2 |
AK052486.1 | 3 | 329 | ||
AK078974.1 | 3 | 337 | ||
AK170817.1 | 3 | 327 | BAE42049.1 |
