Assay Details
Target Gene Details
Entrez Gene ID: | 19739 |
Gene Name: | regulator of G-protein signaling 9 |
Gene Aliases: |
RGS9-1, Rgs9-2 |
Location: |
Chr.11:109225348-109298181 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs9 | NM_001165934.1 | NP_001159406.1 | ||
NM_011268.2 | NP_035398.2 | |||
AF011358.1 | AAC99481.1 | |||
AF125046.1 | AAD20014.1 | |||
AK085443.1 | ||||
AK138159.1 | BAE23565.1 | |||
AK164961.1 | ||||
BC129898.1 | ||||
BC129899.1 |
