Assay Details
Target Gene Details
Entrez Gene ID: | 380683 |
Gene Name: | SEC14-like 3 (S. cerevisiae) |
Gene Aliases: |
1110069O07Rik |
Location: |
Chr.11:4064853-4078990 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sec14l3 | NM_001029937.2 | NP_001025108.1 | ||
BC138982.1 | ||||
BC138985.1 |
