Assay Details
Target Gene Details
Entrez Gene ID: | 110809 |
Gene Name: | serine/arginine-rich splicing factor 1 |
Gene Aliases: |
1110054N12Rik, 5730507C05Rik, 6330415C05Rik, AI482334, AW491331, Asf, Sf2, Sfrs1 |
Location: |
Chr.11:88046902-88053757 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf1 | NM_001078167.2 | 3 | 928 | NP_001071635.1 |
| NM_173374.4 | 3 | 928 | NP_775550.2 | |
| AK018176.1 | 3 | 597 | BAC25546.1 | |
| AK053674.1 | 3 | 619 | ||
| AK078715.1 | 3 | 585 | BAC37367.1 | |
| AK150535.1 | 3 | 631 | BAE29641.1 | |
| AK169203.1 | 3 | 585 | ||
| AK169623.1 | 3 | 586 | ||
| AK187083.1 | ||||
| BC058627.1 | 3 | 594 | AAH58627.1 | |
| BY743476.1 | ||||
| X66091.1 | 3 | 541 |
More Information
Set Membership: |
|

Genomic Map