Assay Details
Target Gene Details
Entrez Gene ID: | 56013 |
Gene Name: | SRC kinase signaling inhibitor 1 |
Gene Aliases: |
P140, SNIP, mKIAA1684, p140Cap |
Location: |
Chr.11:97509339-97575126 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srcin1 | NM_018873.2 | 16 | 3050 | NP_061361.2 |
| AF040944.1 | 16 | 3052 | AAC15635.1 | |
| AK129422.1 | 11 | 2207 | BAC98232.1 | |
| U59873.1 | 16 | 3051 | AAD00087.1 |

Genomic Map