Assay Details
Target Gene Details
Entrez Gene ID: | 68585 |
Gene Name: | reticulon 4 |
Gene Aliases: |
1110020G17Rik, AA407876, AA409940, AA960376, ASY, C130026I10Rik, NOGO, NSP-CL, NgA, Nogo-A, Nogo-B, Nogo-C, mKIAA0886, mKIAA4153 |
Location: |
Chr.11:29692898-29744414 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtn4 | NM_194051.3 | NP_918940.1 | ||
| NM_194052.3 | NP_918941.1 | |||
| NM_194053.3 | NP_918942.1 | |||
| NM_194054.3 | NP_918943.1 | |||
| AK034902.1 | ||||
| AK211699.1 | ||||
| AK218123.1 | ||||
| AK220511.1 | BAD90301.1 | |||
| AY102280.1 | AAM73502.1 | |||
| AY102281.1 | AAM73503.1 | |||
| AY102282.1 | AAM73504.1 | |||
| AY102284.1 | AAM73506.1 | |||
| AY114152.1 | AAM77068.1 | |||
| AY114153.1 | AAM77069.1 |
More Information
Set Membership: |
|

Genomic Map