Assay Details
Target Gene Details
Entrez Gene ID: | 68585 |
Gene Name: | reticulon 4 |
Gene Aliases: |
1110020G17Rik, AA407876, AA409940, AA960376, ASY, C130026I10Rik, NOGO, NSP-CL, NgA, Nogo-A, Nogo-B, Nogo-C, mKIAA0886, mKIAA4153 |
Location: |
Chr.11:29692898-29744414 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtn4 | NM_024226.4 | 2 | 377 | NP_077188.1 |
| NM_194051.3 | 4 | 2664 | NP_918940.1 | |
| NM_194052.3 | 2 | 853 | NP_918941.1 | |
| NM_194053.3 | 3 | 910 | NP_918942.1 | |
| NM_194054.3 | 4 | 3271 | NP_918943.1 | |
| AB073672.1 | 2 | 1229 | BAC75974.1 | |
| AF326337.1 | 2 | 157 | AAK08076.1 | |
| AK034902.1 | 2 | 797 | ||
| AK211699.1 | 2 | 93 | ||
| AK212390.1 | 2 | 80 | ||
| AK218123.1 | 2 | 82 | ||
| AK220511.1 | 4 | 3231 | BAD90301.1 | |
| AY102280.1 | 4 | 2664 | AAM73502.1 | |
| AY102281.1 | 2 | 810 | AAM73503.1 | |
| AY102282.1 | 3 | 867 | AAM73504.1 | |
| AY102283.1 | 2 | 370 | AAM73505.1 | |
| AY102284.1 | 4 | 3228 | AAM73506.1 | |
| AY114152.1 | 4 | 3226 | AAM77068.1 | |
| AY114153.1 | 3 | 681 | AAM77069.1 | |
| BC032192.1 | 2 | 1523 | AAH32192.1 | |
| BU848611.1 | 2 | 216 | ||
| BY251875.1 | 2 | 377 |

Genomic Map