Assay Details
Target Gene Details
Entrez Gene ID: | 380683 |
Gene Name: | SEC14-like 3 (S. cerevisiae) |
Gene Aliases: |
1110069O07Rik |
Location: |
Chr.11:4064853-4078990 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sec14l3 | NM_001029937.2 | 9 | 811 | NP_001025108.1 |
AK136060.1 | 3 | 164 | BAE22802.1 | |
BC138982.1 | 9 | 811 | ||
BC138985.1 | 9 | 811 |
