Assay Details
Target Gene Details
Entrez Gene ID: | 24052 |
Gene Name: | sarcoglycan, delta (dystrophin-associated glycoprotein) |
Gene Aliases: |
35kDa, delta-SG |
Location: |
Chr.11:46971077-47988385 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgcd | NM_011891.4 | NP_036021.1 | ||
| AB024923.1 | BAA83494.1 | |||
| AK040974.1 | BAC30766.1 | |||
| BC130019.1 |
More Information
Set Membership: |
|

Genomic Map