Assay Details
Target Gene Details
Entrez Gene ID: | 327978 |
Gene Name: | schlafen 5 |
Gene Aliases: |
AI956840 |
Location: |
Chr.11:82951349-82964850 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slfn5 | NM_183201.4 | NP_899024.3 | ||
| AK036486.1 | BAC29447.1 | |||
| AK039815.1 | BAE20555.1 | |||
| AK040359.1 | BAE20578.1 | |||
| AK171886.1 | BAE42719.1 |
Target Gene Details
Entrez Gene ID: | 76392 |
Gene Name: | schlafen 5, opposite strand |
Gene Aliases: |
1700003F17Rik |
Location: |
Chr.11:82942341-82960681 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slfn5os | NR_045932.1 | |||
| AK005634.1 | BAB24160.1 | |||
| BY705829.1 |
More Information
Set Membership: |
|

Genomic Map