Assay Details
Target Gene Details
Entrez Gene ID: | 19364 |
Gene Name: | RAD51 homolog D |
Gene Aliases: |
R51H3, Rad51l3, TRAD |
Location: |
Chr.11:82871960-82890751 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rad51d | NM_001277938.1 | 9 | 1017 | NP_001264867.1 |
NM_001277939.1 | 9 | 1032 | NP_001264868.1 | |
NM_001277941.1 | 8 | 897 | NP_001264870.1 | |
NM_001277942.1 | 7 | 870 | NP_001264871.1 | |
NM_011235.4 | 9 | 1032 | NP_035365.1 | |
NR_102717.1 | 8 | 961 | ||
NR_102718.1 | 9 | 1055 | ||
NR_102719.1 | 8 | 913 | ||
NR_102720.1 | 8 | 898 | ||
AB007040.1 | 9 | 997 | BAA24010.1 | |
AB052828.1 | 8 | 926 | BAB19978.1 | |
AB052829.1 | 9 | 982 | BAB19979.1 | |
AB052830.1 | 7 | 835 | BAB19980.1 | |
AB052831.1 | 8 | 878 | BAB19981.1 | |
AB052832.1 | 8 | 863 | BAB19982.1 | |
AB052833.1 | 8 | 862 | BAB19983.1 | |
AB052834.1 | 9 | 1020 | BAB19984.1 | |
AF034955.1 | 9 | 1022 | AAC40093.1 | |
AK017835.1 | 9 | 1015 | BAB30965.1 | |
AK036272.1 | 8 | 871 | ||
AK138553.1 | 9 | 1024 | BAE23698.1 | |
AK147759.1 | 9 | 1007 | BAE28119.1 | |
AK180894.1 | 2 | 113 | ||
CF746344.1 | 5 | 449 | ||
DQ565713.1 | ||||
DQ709189.1 | ||||
EU627688.1 | 9 | 1022 | ||
Y15570.1 | 9 | 1018 | CAA75679.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
