Assay Details
Target Gene Details
Entrez Gene ID: | 56013 |
Gene Name: | SRC kinase signaling inhibitor 1 |
Gene Aliases: |
P140, SNIP, mKIAA1684, p140Cap |
Location: |
Chr.11:97509339-97575126 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Srcin1 | NM_018873.2 | 18 | 3478 | NP_061361.2 |
AF040944.1 | 18 | 3480 | AAC15635.1 | |
AK129422.1 | 13 | 2635 | BAC98232.1 | |
U59873.1 | 18 | 3479 | AAD00087.1 |
