Assay Details
Target Gene Details
Entrez Gene ID: | 103655 |
Gene Name: | SEC14-like 4 (S. cerevisiae) |
Gene Aliases: |
AI256582, AW536553, SPF, TAP |
Location: |
Chr.11:4031378-4052571 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec14l4 | NM_146013.1 | NP_666125.1 | ||
| AK028954.1 | ||||
| BC026948.1 | AAH26948.1 |

Genomic Map