Assay Details
Target Gene Details
Entrez Gene ID: | 237625 |
Gene Name: | phospholipase A2, group III |
Gene Aliases: |
- |
Location: |
Chr.11:3487596-3499346 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g3 | NM_172791.2 | NP_766379.2 | ||
| AK033572.1 | BAC28367.1 | |||
| AK046215.1 | BAC32640.1 | |||
| AK081075.1 | BAC38128.1 | |||
| BC079556.1 | AAH79556.1 |

Genomic Map