Assay Details
Target Gene Details
Entrez Gene ID: | 19084 |
Gene Name: | protein kinase, cAMP dependent regulatory, type I, alpha |
Gene Aliases: |
1300018C22Rik, RIalpha, Tse-1, Tse1 |
Location: |
Chr.11:109649395-109669659 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkar1a | NM_021880.2 | 2 | 256 | NP_068680.1 |
| AK005039.1 | 2 | 258 | BAB23766.1 | |
| AK027916.1 | 2 | 223 | BAC25664.1 | |
| AK049832.1 | 2 | 249 | ||
| AK051068.1 | 2 | 257 | BAC34516.1 | |
| AK053918.1 | 2 | 257 | ||
| AK083383.1 | 2 | 237 | BAC38895.1 | |
| AK145797.1 | 2 | 257 | BAE26655.1 | |
| AK145860.1 | 2 | 224 | BAE26704.1 | |
| AK146380.1 | 2 | 243 | ||
| AK147188.1 | 2 | 248 | BAE27748.1 | |
| AK150427.1 | 2 | 249 | BAE29550.1 | |
| AK151124.1 | 2 | 215 | BAE30132.1 | |
| AK153227.1 | 2 | 215 | BAE31820.1 | |
| AK158562.1 | 2 | 594 | BAE34559.1 | |
| BC003461.1 | 2 | 227 | AAH03461.1 | |
| BC005697.1 | 2 | 198 | AAH05697.1 | |
| HQ412666.1 | 3 | 439 | ||
| HQ412667.1 | 2 | 255 | ||
| HQ412668.1 | 2 | 315 |
More Information
Set Membership: |
|

Genomic Map