Assay Details
Target Gene Details
Entrez Gene ID: | 20528 |
Gene Name: | solute carrier family 2 (facilitated glucose transporter), member 4 |
Gene Aliases: |
Glut-4, Glut4 |
Location: |
Chr.11:69942286-69948221 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc2a4 | NM_009204.2 | NP_033230.2 | ||
| AB008453.1 | BAB03251.1 | |||
| AK137607.1 | BAE23429.1 | |||
| AK138832.1 | ||||
| AK160882.1 | ||||
| AK164310.1 | BAE37730.1 | |||
| BC014282.1 | AAH14282.1 | |||
| M23383.1 | AAA37753.1 |

Genomic Map