Assay Details
Target Gene Details
Entrez Gene ID: | 30939 |
Gene Name: | pituitary tumor-transforming gene 1 |
Gene Aliases: |
AW555095, C87862, Pttg, Pttg3 |
Location: |
Chr.11:43420248-43426248 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pttg1 | NM_001131054.1 | 5 | 508 | NP_001124526.1 |
| NM_013917.2 | 4 | 760 | NP_038945.2 | |
| AF069051.1 | 4 | 744 | AAC62954.1 | |
| AF071209.1 | 4 | 760 | AAC83236.1 | |
| AK002473.1 | 4 | 474 | BAB22127.1 | |
| AK008704.1 | 4 | 475 | BAB25844.1 | |
| AK088387.1 | 4 | 474 | BAC40321.1 | |
| BC023324.1 | 5 | 514 | AAH23324.1 | |
| CN837307.1 | ||||
| DQ157177.1 | 4 | 449 | AAZ83265.1 | |
| DV657646.1 | 4 | 478 |

Genomic Map