Assay Details
Target Gene Details
Entrez Gene ID: | 97761 |
Gene Name: | small G protein signaling modulator 2 |
Gene Aliases: |
D630003G22Rik, R74628, Rutbc1, mKIAA0397 |
Location: |
Chr.11:74849262-74897093 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgsm2 | NM_197943.2 | NP_922934.2 | ||
| AK122273.1 | BAC65555.1 | |||
| AK199501.1 | ||||
| AK202687.1 | ||||
| AK208574.1 | ||||
| BC058414.1 | AAH58414.1 | |||
| BC060163.1 | AAH60163.1 |
More Information
Set Membership: |
|

Genomic Map