Assay Details
Target Gene Details
Entrez Gene ID: | 110172 |
Gene Name: | solute carrier family 35, member B1 |
Gene Aliases: |
Ugalt2 |
Location: |
Chr.11:95384891-95391776 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35b1 | NM_016752.1 | 3 | 358 | NP_058032.1 |
| AK002961.1 | BAC25013.1 | |||
| AK007711.1 | ||||
| AK160124.1 | 3 | 424 | ||
| AK183623.1 | ||||
| AK184790.1 | ||||
| AK185566.1 | ||||
| BC002029.1 | 3 | 339 | AAH02029.1 | |
| D87990.1 | 3 | 358 | BAA13526.1 |
More Information
Set Membership: |
|

Genomic Map