Assay Details
Target Gene Details
Entrez Gene ID: | 11855 |
Gene Name: | Rho GTPase activating protein 5 |
Gene Aliases: |
p190-B, p190B |
Location: |
Chr.12:52503771-52571978 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap5 | NM_009706.2 | NP_033836.2 | ||
| AK131622.1 | BAE20727.1 | |||
| BC150823.1 | ||||
| CF742308.1 | ||||
| CO433888.1 |
More Information
Set Membership: |
|

Genomic Map