Assay Details
Target Gene Details
Entrez Gene ID: | 20747 |
Gene Name: | speckle-type POZ protein |
Gene Aliases: |
AI315626, Pcif1, TEF2 |
Location: |
Chr.11:95414064-95493410 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spop | NM_025287.2 | NP_079563.2 | ||
| AK004669.1 | BAB23458.1 | |||
| AK028201.1 | BAC25809.1 | |||
| AK030746.1 | BAC27114.1 | |||
| AK132383.1 | ||||
| AK159482.1 | BAE35119.1 | |||
| AK160858.1 | ||||
| AK166581.1 | BAE38870.1 | |||
| AK168185.1 | BAE40145.1 | |||
| AK168546.1 | ||||
| BC043131.1 | AAH43131.1 | |||
| BC045205.1 | AAH45205.1 | |||
| BY303667.1 | ||||
| BY726358.1 |
More Information
Set Membership: |
|

Genomic Map