Assay Details
Target Gene Details
Entrez Gene ID: | 20747 |
Gene Name: | speckle-type POZ protein |
Gene Aliases: |
AI315626, Pcif1, TEF2 |
Location: |
Chr.11:95414064-95493410 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spop | NM_025287.2 | 3 | 195 | NP_079563.2 |
| AB071989.1 | BAB68542.1 | |||
| AK004669.1 | 3 | 165 | BAB23458.1 | |
| AK028201.1 | 2 | 331 | BAC25809.1 | |
| AK030746.1 | 2 | 382 | BAC27114.1 | |
| AK132383.1 | 2 | 371 | ||
| AK159482.1 | 2 | 348 | BAE35119.1 | |
| AK160858.1 | 2 | 291 | ||
| AK166581.1 | 2 | 305 | BAE38870.1 | |
| AK168185.1 | 2 | 317 | BAE40145.1 | |
| AK168546.1 | 2 | 384 | ||
| AK216748.1 | ||||
| AY538613.1 | AAT08952.1 | |||
| BC043131.1 | 2 | 214 | AAH43131.1 | |
| BC045205.1 | 3 | 165 | AAH45205.1 | |
| BY303667.1 | 3 | 200 | ||
| BY726358.1 | 3 | 196 |
More Information
Set Membership: |
|

Genomic Map