Assay Details
Target Gene Details
Entrez Gene ID: | 103694 |
Gene Name: | transmembrane emp24 protein transport domain containing 4 |
Gene Aliases: |
1110014L17Rik, AI326346 |
Location: |
Chr.11:6270714-6274837 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmed4 | NM_134020.1 | NP_598781.1 | ||
| AF368921.1 | AAM20821.1 | |||
| AK078425.1 | BAC37268.1 | |||
| AK158876.1 | BAE34708.1 | |||
| AK182125.1 | ||||
| BC023041.1 | AAH23041.1 |
More Information
Set Membership: |
|

Genomic Map