Assay Details
Target Gene Details
Entrez Gene ID: | 72873 |
Gene Name: | coiled-coil domain containing 176 |
Gene Aliases: |
2900006K08Rik, AI427784 |
Location: |
Chr.12:84409068-84441763 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc176 | NM_028377.3 | 2 | 265 | NP_082653.2 |
| AK013494.1 | 2 | 266 | BAB28880.1 | |
| AK133381.1 | 2 | 220 | BAE21625.1 | |
| BC151099.1 | 2 | 187 | ||
| BC151103.1 | 2 | 187 |

Genomic Map