Assay Details
Target Gene Details
Entrez Gene ID: | 108903 |
Gene Name: | tubulin-specific chaperone d |
Gene Aliases: |
2310057L06Rik, A030005L14Rik, mKIAA0988 |
Location: |
Chr.11:121450584-121617170 on Build GRCm38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbcd | NM_029878.3 | NP_084154.1 | ||
| AK041464.1 | BAC30951.1 | |||
| AK050199.1 | ||||
| AK054343.1 | ||||
| AK185941.1 | ||||
| BC059843.1 | AAH59843.1 |
Target Gene Details
Entrez Gene ID: | 319530 |
Gene Name: | zinc finger protein 750 |
Gene Aliases: |
A030007D23Rik, Znf750 |
Location: |
Chr.11:121510976-121519342 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp750 | NM_178763.4 | 1 | 60 | NP_848878.1 |
| AK028879.1 | ||||
| AK030690.1 | BAC27080.1 | |||
| AK037185.1 | BAC29743.1 | |||
| AK037240.1 | BAC29771.1 | |||
| AK076268.1 | 1 | 52 | ||
| CA555996.1 | 1 | 60 |
More Information
Set Membership: |
|

Genomic Map