Assay Details
Target Gene Details
Entrez Gene ID: | 68764 |
Gene Name: | cadherin-related family member 3 |
Gene Aliases: |
1110049B09Rik, BB172694, Cdh28 |
Location: |
Chr.12:33033796-33093037 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdhr3 | NM_001024478.1 | 2 | 214 | NP_001019649.1 |
| AK047666.1 | 2 | 215 | BAC33118.1 | |
| AK049806.1 | 2 | 216 | ||
| BC138911.1 | 2 | 142 |
More Information
Set Membership: |
|

Genomic Map