Assay Details
Target Gene Details
Entrez Gene ID: | 212073 |
Gene Name: | spectrin repeat containing, nuclear envelope family member 3 |
Gene Aliases: |
nesprin-3 |
Location: |
Chr.12:104929830-105010098 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syne3 | NM_001042699.2 | 6 | 1002 | NP_001036164.1 |
| NM_172500.3 | 4 | 647 | NP_766088.2 | |
| AK029216.1 | 4 | 649 | BAC26351.1 | |
| BC099694.1 | 6 | 1002 | AAH99694.1 |

Genomic Map