Assay Details
Target Gene Details
Entrez Gene ID: | 217337 |
Gene Name: | signal recognition particle 68 |
Gene Aliases: |
2610024I03Rik |
Location: |
Chr.11:116245166-116274217 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srp68 | NM_146032.3 | NP_666144.3 | ||
| AK033010.1 | BAC28124.1 | |||
| AK076003.1 | ||||
| AK180834.1 | ||||
| AK199914.1 | ||||
| BC033573.1 | AAH33573.1 | |||
| BC070422.1 | AAH70422.1 |
More Information
Set Membership: |
|

Genomic Map