Assay Details
Target Gene Details
Entrez Gene ID: | 217337 |
Gene Name: | signal recognition particle 68 |
Gene Aliases: |
2610024I03Rik |
Location: |
Chr.11:116245166-116274217 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srp68 | NM_146032.3 | 9 | 1139 | NP_666144.3 |
| AK033010.1 | 9 | 1058 | BAC28124.1 | |
| AK076003.1 | 9 | 1036 | ||
| AK187819.1 | 1 | 66 | ||
| AK199914.1 | 4 | 299 | ||
| BC033573.1 | 5 | 439 | AAH33573.1 | |
| BC070422.1 | 9 | 1139 | AAH70422.1 |
More Information
Set Membership: |
|

Genomic Map