Assay Details
Target Gene Details
Entrez Gene ID: | 20868 |
Gene Name: | serine/threonine kinase 10 |
Gene Aliases: |
Gek1, Lok, mKIAA4026 |
Location: |
Chr.11:32533266-32624595 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk10 | NM_009288.2 | NP_033314.2 | ||
| AK148118.1 | BAE28356.1 | |||
| AK220448.1 | BAD90480.1 | |||
| BC128363.1 | ||||
| D89728.1 | BAA24073.1 |
More Information
Set Membership: |
|

Genomic Map