Assay Details
Target Gene Details
Entrez Gene ID: | 21681 |
Gene Name: | Aly/REF export factor |
Gene Aliases: |
ALY, REF1, Refbp1, Thoc4 |
Location: |
Chr.11:120594516-120598436 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alyref | NM_011568.1 | NP_035698.1 | ||
| AJ252140.1 | CAB76383.1 | |||
| BC120588.1 | ||||
| BC137658.1 | ||||
| U89876.1 | AAC53117.1 |
More Information
Set Membership: |
|

Genomic Map